1,4-Dithioerythritol ≥99%, Molecular Biology Grade
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
DTE
BI8765-10G
BI8765-25G
BI8765-5G
APOSBI8765-10GEA
3310
CZK
APOSBI8765-10G
APOSBI8765-25G
APOSBI8765-5G
1,4-Dithioerythritol ≥99%, Molecular Biology Grade
1,4-Dithioerythritol
Reagent for maintaining -SH groups in reduced state.
Vzorec:
C₄H₁₀O₂S₂ MW: 154,25 g/mol Bod tání: 82…84 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063750 CAS číslo: 6892-68-8 EINECS: 229-998-8 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...