Triphenylmethyl mercaptan 98%
Dodavatel: Apollo Scientific
Synonyms:
Triphenylmethanethiol
BI7573-25G
BI7573-100G
APOSBI7573-25GEA
1580
CZK
APOSBI7573-25G
APOSBI7573-100G
Triphenylmethyl mercaptan 98%
Triphenylmethyl mercaptan
Vzorec:
C₁₉H₁₆S MW: 276,40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004854 CAS číslo: 3695-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...