Tisk…

2-Amino-2-methyl-1,3-propanediol 99+%, high purity

Dodavatel: Apollo Scientific
Varování

BI4545-100G BI4545-25G
APOSBI4545-100GEA 2150 CZK
APOSBI4545-100G APOSBI4545-25G
2-Amino-2-methyl-1,3-propanediol 99+%, high purity
2-Amino-2-methyl-1,3-propanediol

Buffer component used in SDS - gradient gel electrophoresis. Biological buffer - useful range 7.8-9.7.


  • White crystals
  • Water (by Karl Fischer) <1.0%
  • Forms a clear solution in water.

Vzorec: C₄H₁₁NO₂
MW: 105,14 g/mol
Bod varu: 151 °C/10mmHg
Bod tání: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004678
CAS číslo: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR