L-(-)-Fucose
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal
BI3933-10G
BI3933-25G
BI3933-50G
APOSBI3933-10GEA
3740
CZK
APOSBI3933-10G
APOSBI3933-25G
APOSBI3933-50G
L-(-)-Fucose
L(-)-Fucose
Vzorec:
C₆H₁₂O₅ MW: 164,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069812 CAS číslo: 2438-80-4 EINECS: 219-452-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...