Fluconazole, anhydrous
Dodavatel: Apollo Scientific
|
Varování
|
BI3858-1G
BI3858-5G
BI3858-25G
APOSBI3858-1GEA
622
CZK
APOSBI3858-1G
APOSBI3858-5G
APOSBI3858-25G
Fluconazole, anhydrous
Fluconazole
Vzorec:
C₁₃H₁₂F₂N₆O Storage Temperature: Ambient |
MDL Number:
MFCD00274549 CAS číslo: 86386-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...