Tisk…
Tris-(hydroxymethyl)aminomethan ≥99.9%, Ultrapure
Dodavatel: Apollo Scientific
Varování
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
BI2888-1KG BI2888-500G BI2888-5KG
APOSBI2888-1KGEA 1580 CZK
APOSBI2888-1KG APOSBI2888-500G APOSBI2888-5KG
Tris-(hydroxymethyl)aminomethan ≥99.9%, Ultrapure
Tris-(hydroxymethyl)aminomethan
  • Loss on drying: <0.2%
  • Heavy metals: <1 ppm
  • pka: @ 20°C:8.3 + 0.1
  • Biological buffer - useful range 7.0 - 9.0
Vzorec: NH₂C(CH₂OH)₃
MW: 121.14
Bod varu: 219…220 °C
Bod tání: 171.2…172.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS číslo: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842