Tisk…

Mycophenolic acid

Dodavatel: Apollo Scientific
Nebezpečí

BI1535-250MG BI1535-100MG
APOSBI1535-250MGEA 3020 CZK
APOSBI1535-250MG APOSBI1535-100MG
Mycophenolic acid
Mycophenolic acid

Immunosuppressive agent.

Vzorec: C₁₇H₂₀O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS číslo: 24280-93-1
EINECS: 246-119-3
UN: 3077
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR