Tisk…

2-Acetamido-3-fluorobenzoic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(Acetylamino)-3-fluorobenzoic acid, N-(2-Carboxy-6-fluorophenyl)acetamide, N-Acetyl-3-fluoroanthranilic acid

PC9979-1G PC9979-500MG
APOSPC9979-1GEA 14880 CZK
APOSPC9979-1G APOSPC9979-500MG
2-Acetamido-3-fluorobenzoic acid
2-Acetamido-3-fluorobenzoic acid
Vzorec: C₉H₈FNO₃
MW: 197,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094569
CAS číslo: 550346-18-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR