Ethyl 3,5-difluoro-4-methylbenzoate 95%
Dodavatel: Apollo Scientific
PC99023-1G
PC99023-500MG
APOSPC99023-1GEA
7870
CZK
APOSPC99023-1G
APOSPC99023-500MG
Ethyl 3,5-difluoro-4-methylbenzoate 95%
Ethyl 3,5-difluoro-4-methylbenzoate
Vzorec:
C₁₀H₁₀F₂O₂ MW: 200,182 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28790312 CAS číslo: 1806321-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...