Tisk…

3-Bromo-2-chloro-6-fluorotoluene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-2-chloro-4-fluoro-3-methylbenzene

PC9893-25G PC9893-5G PC9893-100G
APOSPC9893-25GEA 2780 CZK
APOSPC9893-25G APOSPC9893-5G APOSPC9893-100G
3-Bromo-2-chloro-6-fluorotoluene 97%
3-Bromo-2-chloro-6-fluorotoluene
Vzorec: C₇H₅BrClF
MW: 223,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236591
CAS číslo: 203302-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR