Tisk…

4-Hydroxy-2-(pyrid-3-yl)-6-trifluoromethylpyrimidine

Dodavatel: Apollo Scientific

PC9875-1G PC9875-500MG
APOSPC9875-1GEA 9530 CZK
APOSPC9875-1G APOSPC9875-500MG
4-Hydroxy-2-(pyrid-3-yl)-6-trifluoromethylpyrimidine
4-Hydroxy-2-(pyrid-3-yl)-6-trifluoromethylpyrimidine
Vzorec: C₁₀H₆F₃N₃O
MW: 241,169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140055
CAS číslo: 204394-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR