Tisk…

Perfluorodiglyme 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Perfluoro(diethylene glycol dimethyl ether)

PC9776-10G PC9776-5G PC9776-1G
APOSPC9776-10GEA 3510 CZK
APOSPC9776-10G APOSPC9776-5G APOSPC9776-1G
Perfluorodiglyme 98%
Perfluorodiglyme
Vzorec: C₆F₁₄O₃
MW: 386.04 g/mol
Bod varu: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458175
CAS číslo: 40891-99-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR