2,4,4,4-Tetrafluorobut-1-ene 97%
Dodavatel: Apollo Scientific
PC9729-25G
PC9729-5G
APOSPC9729-25GEA
27310
CZK
APOSPC9729-25G
APOSPC9729-5G
2,4,4,4-Tetrafluorobut-1-ene 97%
2,4,4,4-Tetrafluorobut-1-ene
Vzorec:
C₄H₄F₄ MW: 128,068 g/mol Bod varu: 29…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784201 CAS číslo: 721946-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...