Perfluoropentanoyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Nonafluoropentanoyl chloride, Nonafluorovaleryl chloride
PC9708-1G
PC9708-25G
PC9708-5G
APOSPC9708-1GEA
5630
CZK
APOSPC9708-1G
APOSPC9708-25G
APOSPC9708-5G
Perfluoropentanoyl chloride 97%
Perfluoropentanoyl chloride
Vzorec:
C₅ClF₉O Bod varu: 67,5…68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00155890 CAS číslo: 375-60-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...