Tisk…

tert-Butyl chloro(difluoro)acetate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Chloro(difluoro)acetic acid tert-butyl ester, tert-Butyl 2-chloro-2,2-difluoroethanoate

PC9651-25G PC9651-5G
APOSPC9651-25GEA 10090 CZK
APOSPC9651-25G APOSPC9651-5G
tert-Butyl chloro(difluoro)acetate 95%
tert-Butyl chloro(difluoro)acetate
Vzorec: C₆H₉ClF₂O₂
MW: 186,584 g/mol
Bod varu: 65…70 °C (10 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00153088
CAS číslo: 167308-43-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR