Tisk…

5-(4-tert-Butylphenyl)-4-[3-(trifluoromethyl)phenyl]-1,2,4-triazole-3-thiol

Dodavatel: Apollo Scientific

PC9597-1G PC9597-5G PC9597-10G
APOSPC9597-1GEA 2690 CZK
APOSPC9597-1G APOSPC9597-5G APOSPC9597-10G
5-(4-tert-Butylphenyl)-4-[3-(trifluoromethyl)phenyl]-1,2,4-triazole-3-thiol
5-(4-tert-Butylphenyl)-4-[3-(trifluoromethyl)phenyl]-1,2,4-triazole-3-thiol
Vzorec: C₁₉H₁₈F₃N₃S
MW: 377,427 g/mol
Bod tání: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00177364
CAS číslo: 261633-26-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR