Tisk…

5-Chloro-4-fluoroindole 98%

Dodavatel: Apollo Scientific

Synonyms: 5-Chloro-4-fluoro-1H-indole

PC9566-100MG PC9566-1G PC9566-250MG
APOSPC9566-100MGEA 2400 CZK
APOSPC9566-100MG APOSPC9566-1G APOSPC9566-250MG
5-Chloro-4-fluoroindole 98%
5-Chloro-4-fluoroindole
Vzorec: C₈H₅ClFN
MW: 169,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01074494
CAS číslo: 376646-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR