3-Chloro-4-fluorobenzyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-4-(chloromethyl)-1-fluorobenzene, α,3-Dichloro-4-fluorotoluene
PC9555-1G
PC9555-25G
PC9555-5G
APOSPC9555-1GEA
622
CZK
APOSPC9555-1G
APOSPC9555-25G
APOSPC9555-5G
3-Chloro-4-fluorobenzyl chloride 98%
3-Chloro-4-fluorobenzyl chloride
Vzorec:
C₇H₅Cl₂F Storage Temperature: Ambient |
MDL Number:
MFCD03701044 CAS číslo: 2994-69-6 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...