Tisk…

[1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenylthio]-1H-pyrazol-4-yl]methanol 97%

Dodavatel: Apollo Scientific

PC9439-1G PC9439-500MG
APOSPC9439-1GEA 33400 CZK
APOSPC9439-1G APOSPC9439-500MG
[1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenylthio]-1H-pyrazol-4-yl]methanol 97%
[1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenylthio]-1H-pyrazol-4-yl]methanol
Vzorec: C₁₈H₁₅F₃N₂OS
MW: 364,385 g/mol
Bod tání: 134…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00244951
CAS číslo: 318247-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR