Tisk…

3,4-Difluorophenylacetyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC9314-1G PC9314-250MG
APOSPC9314-1GEA 5360 CZK
APOSPC9314-1G APOSPC9314-250MG
3,4-Difluorophenylacetyl chloride
3,4-Difluorophenylacetyl chloride
Vzorec: C₈H₅ClF₂O
MW: 190,575 g/mol
Bod varu: 91…92 °C (6 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08458138
CAS číslo: 121639-61-0
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR