Tisk…

2-Fluoro-6-methylbenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Bromomethyl)-1-fluoro-3-methylbenzene, α'-Bromo-3-fluoro-o-xylene

PC9288-1G PC9288-5G
APOSPC9288-1GEA 5460 CZK
APOSPC9288-1G APOSPC9288-5G
2-Fluoro-6-methylbenzyl bromide
2-Fluoro-6-methylbenzyl bromide
Vzorec: C₈H₈BrF
MW: 203.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660281
CAS číslo: 886502-18-7
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR