2-(Aminomethyl)-5-(trifluoromethyl)furan 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
[5-(Trifluoromethyl)fur-2-yl]methylamine, 5-(Trifluoromethyl)furfurylamine
PC9250-1G
PC9250-250MG
APOSPC9250-1GEA
16340
CZK
APOSPC9250-1G
APOSPC9250-250MG
2-(Aminomethyl)-5-(trifluoromethyl)furan 97%
2-(Aminomethyl)-5-(trifluoromethyl)furan
Vzorec:
C₆H₆F₃NO MW: 165,113 g/mol Bod varu: 90…100 °C (20 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03086222 CAS číslo: 868755-68-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...