1,6-Dichloroperfluorohexane 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,6-Dichloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
PC9238-10G
PC9238-25G
PC9238-5G
APOSPC9238-10GEA
8300
CZK
APOSPC9238-10G
APOSPC9238-25G
APOSPC9238-5G
1,6-Dichloroperfluorohexane 97%
1,6-Dichloroperfluorohexane
Vzorec:
C₆Cl₂F₁₂ Bod varu: 113…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155741 CAS číslo: 355-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...