5-Bromo-2-(trifluoromethyl)nicotinic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
5-Bromo-2-(trifluoromethyl)pyridine-3-carboxylic acid
PC9198-1G
PC9198-250MG
APOSPC9198-1GEA
0
CZK
APOSPC9198-1G
APOSPC9198-250MG
5-Bromo-2-(trifluoromethyl)nicotinic acid 97%
5-Bromo-2-(trifluoromethyl)nicotinic acid
Vzorec:
C₇H₃BrF₃NO₂ MW: 270,003 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08741350 CAS číslo: 436799-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...