4-Fluoro-1H-indole-3-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC911409-1G
PC911409-250MG
APOSPC911409-1GEA
16630
CZK
APOSPC911409-1G
APOSPC911409-250MG
4-Fluoro-1H-indole-3-carbonitrile 97%
4-Fluoro-1H-indole-3-carbonitrile
Vzorec:
C₉H₅FN₂ MW: 160,148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15527152 CAS číslo: 1260759-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...