Tisk…

7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 95%

Dodavatel: Apollo Scientific

PC911377-1G PC911377-250MG
APOSPC911377-1GEA 2650 CZK
APOSPC911377-1G APOSPC911377-250MG
7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine 95%
7-(Trifluoromethyl)imidazo[1,2-a]pyrimidine
Vzorec: C₇H₄F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD08234997
CAS číslo: 375857-66-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR