Tisk…

2-Cyano-3-(2-fluorophenyl)propionic acid 95%

Dodavatel: Apollo Scientific

PC911251-1G PC911251-250MG
APOSPC911251-1GEA 21490 CZK
APOSPC911251-1G APOSPC911251-250MG
2-Cyano-3-(2-fluorophenyl)propionic acid 95%
2-Cyano-3-(2-fluorophenyl)propionic acid
Vzorec: C₁₀H₈FNO₂
MW: 193,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800431
CAS číslo: 948015-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR