Tisk…

(R)-b-Amino-4-fluoro-benzeneethanol 97%

Dodavatel: Apollo Scientific

PC911186-1G PC911186-250MG PC911186-500MG
APOSPC911186-1GEA 22320 CZK
APOSPC911186-1G APOSPC911186-250MG APOSPC911186-500MG
(R)-b-Amino-4-fluoro-benzeneethanol 97%
(R)-b-Amino-4-fluoro-benzeneethanol
Vzorec: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD09253715
CAS číslo: 174770-74-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR