Tisk…

4-Nitro-5-trifluoromethyl-1H-pyrazole 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Nitro-5-(Trifluoromethyl)-1H-pyrazole

PC911116-1G PC911116-250MG PC911116-5G
APOSPC911116-1GEA 3970 CZK
APOSPC911116-1G APOSPC911116-250MG APOSPC911116-5G
4-Nitro-5-trifluoromethyl-1H-pyrazole 98%
4-Nitro-5-trifluoromethyl-1H-pyrazole
Vzorec: C₄H₂F₃N₃O₂
MW: 181,073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971110
CAS číslo: 1046462-99-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR