Tisk…

1-Boc-3-(4-fluorobenzyl)-3-carboxypiperidine 97%

Dodavatel: Apollo Scientific

PC911085-1G PC911085-250MG PC911085-500MG
APOSPC911085-1GEA 22320 CZK
APOSPC911085-1G APOSPC911085-250MG APOSPC911085-500MG
1-Boc-3-(4-fluorobenzyl)-3-carboxypiperidine 97%
1-Boc-3-(4-fluorobenzyl)-3-carboxypiperidine
Vzorec: C₁₈H₂₄FNO₄
MW: 337.386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840049
CAS číslo: 887344-22-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR