Tisk…

3-(Difluoromethoxy)-4-hydroxybenzaldehyde 95+%

Dodavatel: Apollo Scientific
Varování

PC911011-100MG PC911011-250MG
APOSPC911011-100MGEA 1540 CZK
APOSPC911011-100MG APOSPC911011-250MG
3-(Difluoromethoxy)-4-hydroxybenzaldehyde 95+%
3-(Difluoromethoxy)-4-hydroxybenzaldehyde
Vzorec: C₈H₆F₂O₃
MW: 188.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00720656
CAS číslo: 53173-70-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR