2-(Difluoromethyl)-5-nitropyridine 95%
Dodavatel: Apollo Scientific
PC910983-1G
PC910983-250MG
APOSPC910983-1GEA
3060
CZK
APOSPC910983-1G
APOSPC910983-250MG
2-(Difluoromethyl)-5-nitropyridine 95%
2-(Difluoromethyl)-5-nitropyridine
Vzorec:
C₆H₄F₂N₂O₂ MW: 174,105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25476575 CAS číslo: 1646152-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...