1-(4-Fluorophenyl)hexan-1-one 98%
Dodavatel: Apollo Scientific
PC910952-1G
PC910952-5G
APOSPC910952-1GEA
7670
CZK
APOSPC910952-1G
APOSPC910952-5G
1-(4-Fluorophenyl)hexan-1-one 98%
1-(4-Fluorophenyl)hexan-1-one
Vzorec:
C₁₂H₁₅FO MW: 194.245 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11210352 CAS číslo: 1426-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...