4-Fluoro-3-(hydroxymethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
PC910950-1G
PC910950-250MG
APOSPC910950-1GEA
16960
CZK
APOSPC910950-1G
APOSPC910950-250MG
4-Fluoro-3-(hydroxymethyl)benzonitrile 98%
4-Fluoro-3-(hydroxymethyl)benzonitrile
Vzorec:
C₈H₆FNO Storage Temperature: Ambient |
MDL Number:
MFCD11213689 CAS číslo: 856931-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...