Tisk…

3-Chloro-1-(4-fluorophenyl)propan-1-ol 96%

Dodavatel: Apollo Scientific

PC910933-1G PC910933-5G
APOSPC910933-1GEA 7670 CZK
APOSPC910933-1G APOSPC910933-5G
3-Chloro-1-(4-fluorophenyl)propan-1-ol 96%
3-Chloro-1-(4-fluorophenyl)propan-1-ol
Vzorec: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD14529705
CAS číslo: 31736-75-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR