Tisk…

[3-(2-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine 98%

Dodavatel: Apollo Scientific

PC910925-1G PC910925-5G
APOSPC910925-1GEA 7050 CZK
APOSPC910925-1G APOSPC910925-5G
[3-(2-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine 98%
[3-(2-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine
Vzorec: C₉H₈FN₃O
MW: 193,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08899396
CAS číslo: 919017-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR