Tisk…

[3-(3-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine 98%

Dodavatel: Apollo Scientific

PC910922-1G PC910922-250MG
APOSPC910922-1GEA 14290 CZK
APOSPC910922-1G APOSPC910922-250MG
[3-(3-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine 98%
[3-(3-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine
Vzorec: C₉H₈FN₃O
MW: 193,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09034253
CAS číslo: 937651-13-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR