N-Isobutyl 3-fluorobenzylamine 98%
Dodavatel: Apollo Scientific
PC910858-1G
PC910858-5G
APOSPC910858-1GEA
6220
CZK
APOSPC910858-1G
APOSPC910858-5G
N-Isobutyl 3-fluorobenzylamine 98%
N-Isobutyl 3-fluorobenzylamine
Vzorec:
C₁₁H₁₆FN MW: 181,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11139645 CAS číslo: 1019578-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...