8-Bromo-6-fluorochroman-4-one 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC910812-100MG
PC910812-250MG
APOSPC910812-100MGEA
1740
CZK
APOSPC910812-100MG
APOSPC910812-250MG
8-Bromo-6-fluorochroman-4-one 97%
8-Bromo-6-fluorochroman-4-one
Vzorec:
C₉H₆BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11104781 CAS číslo: 1092350-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...