Tisk…

Methyl 5-fluoro-2-methoxypyridine-3-carboxylate 96%

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-fluoro-2-methoxynicotinate

PC910765-1G PC910765-250MG
APOSPC910765-1GEA 14880 CZK
APOSPC910765-1G APOSPC910765-250MG
Methyl 5-fluoro-2-methoxypyridine-3-carboxylate 96%
Methyl 5-fluoro-2-methoxypyridine-3-carboxylate
Vzorec: C₈H₈FNO₃
MW: 185,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698160
CAS číslo: 122433-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR