8-Fluoro-4-methyl-1H-quinolin-2-one 98%
Dodavatel: Apollo Scientific
PC910722-1G
PC910722-250MG
APOSPC910722-1GEA
23580
CZK
APOSPC910722-1G
APOSPC910722-250MG
8-Fluoro-4-methyl-1H-quinolin-2-one 98%
8-Fluoro-4-methyl-1H-quinolin-2-one
Vzorec:
C₁₀H₈FNO MW: 177,175 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22373149 CAS číslo: 5279-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...