Tisk…

6-Chloro-2-fluoropyridine-3-carbaldehyde 97%

Dodavatel: Apollo Scientific

Synonyms: 6-Chloro-2-fluoro-3-pyridinecarboxaldehyde, 6-Chloro-2-fluoronicotinaldehyde

PC910712-1G PC910712-250MG
APOSPC910712-1GEA 10060 CZK
APOSPC910712-1G APOSPC910712-250MG
6-Chloro-2-fluoropyridine-3-carbaldehyde 97%
6-Chloro-2-fluoropyridine-3-carbaldehyde
Vzorec: C₆H₃ClFNO
MW: 159,546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16556251
CAS číslo: 1093880-37-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR