Tisk…

3-Bromo-8-fluoroquinolin-6-amine 97%

Dodavatel: Apollo Scientific
Varování

PC910648-1G PC910648-250MG
APOSPC910648-1GEA 6720 CZK
APOSPC910648-1G APOSPC910648-250MG
3-Bromo-8-fluoroquinolin-6-amine 97%
3-Bromo-8-fluoroquinolin-6-amine
Vzorec: C₉H₆BrFN₂
MW: 241.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24539398
CAS číslo: 1432323-09-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR