3-Cyclopropoxy-5-fluoro-N-methylaniline 95%
Dodavatel: Apollo Scientific
PC910627-1G
PC910627-250MG
APOSPC910627-1GEA
5960
CZK
APOSPC910627-1G
APOSPC910627-250MG
3-Cyclopropoxy-5-fluoro-N-methylaniline 95%
3-Cyclopropoxy-5-fluoro-N-methylaniline
Vzorec:
C₁₀H₁₂FNO Storage Temperature: Ambient |
MDL Number:
MFCD17004957 CAS číslo: 1243481-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...