Tisk…

Ethyl 2-fluoro-5-hydroxybenzoate 95%

Dodavatel: Apollo Scientific

PC910289-1G PC910289-5G
APOSPC910289-1GEA 6220 CZK
APOSPC910289-1G APOSPC910289-5G
Ethyl 2-fluoro-5-hydroxybenzoate 95%
Ethyl 2-fluoro-5-hydroxybenzoate
Vzorec: C₉H₉FO₃
MW: 184,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12406844
CAS číslo: 1214387-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR