4-Fluoro-2,5-dimethylphenol 95%
Dodavatel: Apollo Scientific
PC910227-1G
PC910227-250MG
APOSPC910227-1GEA
6290
CZK
APOSPC910227-1G
APOSPC910227-250MG
4-Fluoro-2,5-dimethylphenol 95%
4-Fluoro-2,5-dimethylphenol
Vzorec:
C₈H₉FO Storage Temperature: Ambient |
MDL Number:
MFCD23111064 CAS číslo: 196519-58-1 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...