Tisk…

2-(Difluoromethoxy)pyridin-4-amine 97%

Dodavatel: Apollo Scientific
Varování

PC910171-100MG PC910171-250MG
APOSPC910171-100MGEA 1290 CZK
APOSPC910171-100MG APOSPC910171-250MG
2-(Difluoromethoxy)pyridin-4-amine 97%
2-(Difluoromethoxy)pyridin-4-amine
Vzorec: C₆H₆F₂N₂O
MW: 160,121 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18261290
CAS číslo: 1211587-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR