5-(3-Fluorophenyl)pyridin-2-amine 98%
Dodavatel: Apollo Scientific
PC909622-1G
PC909622-5G
APOSPC909622-1GEA
5390
CZK
APOSPC909622-1G
APOSPC909622-5G
5-(3-Fluorophenyl)pyridin-2-amine 98%
5-(3-Fluorophenyl)pyridin-2-amine
Vzorec:
C₁₁H₉FN₂ MW: 188,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06802132 CAS číslo: 866620-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...