Tisk…

4-Fluoro-1H-benzo[d]imidazol-2(3H)-one 97%

Dodavatel: Apollo Scientific
Varování

PC909355-1G PC909355-250MG
APOSPC909355-1GEA 6050 CZK
APOSPC909355-1G APOSPC909355-250MG
4-Fluoro-1H-benzo[d]imidazol-2(3H)-one 97%
4-Fluoro-1H-benzo[d]imidazol-2(3H)-one
Vzorec: C₇H₅FN₂O
MW: 152,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18814470
CAS číslo: 256519-10-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR