Tisk…

4-Fluoro-2-(methylsulphanyl)-1-nitrobenzene 98%

Dodavatel: Apollo Scientific

PC909184-1G PC909184-5G
APOSPC909184-1GEA 8870 CZK
APOSPC909184-1G APOSPC909184-5G
4-Fluoro-2-(methylsulphanyl)-1-nitrobenzene 98%
4-Fluoro-2-(methylsulphanyl)-1-nitrobenzene
Vzorec: C₇H₆FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD22489274
CAS číslo: 78227-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR